Learn More
MilliporeSigma™ Replacement Rolls for MColorpHast™ Neutralit™ pH Indicator Papers, pH 5.5 to 9.0
Roll (4.8 m) with colour scale pH 5.5 - 6.0 - 6.5 - 7.0 - 7.5 - 8.0 - 8.5 - 9.0 Neutralit™
Manufacturer:  MilliporeSigma™ 1.09564.0003
 View more versions of this product
Catalog No. M1095640003
Add to cart
Description
Description
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences. pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.
Specifications
Specifications
Litmus Paper Replacement Roll
pH indicator papers, 109564, 109565 Neutralit™
3 Rolls
5.5 to 9
For long term storage, since they are protected against external influences
5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0
5.5-6.0-6.5-7.0-7.5-8.0-8.5-9.0 mg/L