Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RiOs™ 30/50/100 Water Purification Systems

For ensuring quality, the Millipore RiOs™ 30/50/100 Water Purification System can serve a single lab, suite of labs, or an entire laboratory building, depending on model.

Specifications

Conductivity <2000μS/cm
Contaminant Rejection (Microbiological) Less than10cfu/mL
Contaminant Rejection (Organic) Greater than99.9%
Depth (English) 17.4 in.
Depth (Metric) 44.1cm
View More Specs

Requires:

  • Requires 8mm tubing for product and Elix module reject water
  • 1/2 in. tubing for RO reject water
  • >11/3 gal./min. (5L/min.) drain


Products 1
Catalog Number Mfr. No. Daily Output Model Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Daily Output Model Weight (English) Weight (Metric) Price Quantity & Availability  
ZR0S60050
View Documents
MilliporeSigma™
ZR0S60050
1000L RiOs™ 50 75 lb. 34kg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Combine water purification, storage, distribution and control in a single unit. Provide hundreds to thousands of L/day of Type III laboratory grade water from tap.

Key Features

  • Utilize a three-step water treatment process
  • Provide total control of all functions, operating parameters and standard accessory components within the RiOs system itself and within the external pure water distribution loop
  • Do not usually require an external control box

Pretreatment Features

  • Progard™ L pack removes particles (0.5µm filter), free chlorine, and colloids (activated carbon filter) from tap water to protect the system
  • Anti-scaling agent protects the reverse osmosis membrane in hard water areas
  • Bactericidal carbon prevents unwanted bacteria growth
  • Pretreatment packs are tailored to the feedwater source
Advanced Reverse Osmosis (RO) Features
  • Removes 95 to 99% of inorganic ions and 99% of all dissolved organics (MW 200 Daltons), microorganisms and particles
  • Recycles part of the RO reject water back to the RO membrane feedwater stream
  • Allows water recovery to be adjusted up to 70% to optimize water consumption, depending on the feedwater quality and the pretreatment sequence used
  • Maintains constant flowrate from 7 to 30°C
Ultraviolet Sanitization Features
  • Uses a 254nm UV lamp
  • Leads to a log reduction value (LRV) of four in the bacteria count of the water, irrespective of the system's nominal flowrate
System Operation Features
  • After each purification step, relevant parameters are checked by the system
  • Water quality monitoring within the pure water distribution loop (resistivity) can also be checked by the system
  • Resistivity test is designed to meet the requirements of USP (645) suitability test
  • Water quality data is recorded by the system in real time for full traceability
  • Solenoid valve feed: 1/2 NPT
  • RS-232 data port connection

  • Produces water ideal for general rinsing, bacteria-sensitive applications, or as feedwater for humidifiers, autoclaves and washing machines
  • Also suitable as a source of feedwater for Milli-Q™ and Super-Q™ ultrapure water systems

†Includes plumbing connections.

Specifications

Specifications

<2000μS/cm
Greater than99.9%
44.1cm
Potable tap water
Water purification system
110V 60Hz
0.5μm
Elix™
Less than2000
7 to 30°C
Less than10cfu/mL
17.4 in.
17.37 x 26.1 x 28.9 in. (44 x 66.2 x 73.4cm)
Less than5
Type 3
2 to 6 bar
60Hz
4 to 10
44.6 to 86°F
110V
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit