Learn More
MilliporeSigma™ Scepter 2.0 Automated Handheld Cytometer Accessory: O-Ring Kit
For use with Scepter 2.0 (see cat. no. PHCC20040)
Manufacturer:  MilliporeSigma™ PHCC0CLIP
Catalog No. PHCC0CLIP
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
After repeated use the Scepter O-ring may wear. It can be easily replaced with this kit and by following the instructions included in the manual.
Specifications
Specifications
O-ring Kit
Counting cells and delivering histrogram profiles of cellular populations
2 O-rings, 1 filter cover
Each
Scepter 2.0 handheld automated cell counter