Learn More
MilliporeSigma™ SeQuant™ SAMS™ and CARS™ Suppressor System: Replacement Cartridges
For use with CARS™ system SeQuant™
Specifications
For Use With SeQuant SAMS and CARS Suppressor System
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
M1506130001
View Documents
MilliporeSigma™
1.50613.0001
Small Replacement cartridge This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
M1506140001
View Documents
MilliporeSigma™
1.50614.0001
Large Replacement cartridge This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
SeQuant™ CARS™ (Continuous Anion Regeneration System) with SAMS™ (SeQuant™ Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This cartridge replaces the one supplied with the CARS™ system when it is depleted.
Specifications
Specifications
SeQuant SAMS and CARS Suppressor System