Learn More
MilliporeSigma™ SNAP i.d.™ 2.0 Protein Detection System: Mini and Midi Blot Holding Frames
Mini and midi blot holding frames for MilliporeSigma SNAP i.d. 2.0 Protein Detection Systems
Manufacturer:  MilliporeSigma™ SNAP2FRMD02
 View more versions of this product
Catalog No. SNAP2FRMD02
Add to cart
Description
Description
For use with EMD Millipore SNAP i.d. 2.0 Protein Detection System (Cat. No. SNAP2MM)
Specifications
Specifications
Midi Blot Holding Frames
Clear lids, easy locking latch, umbrella valve
2/Pk.
Snap i.d.™ 2.0 Protein detection system-midi
Yes