Learn More
MilliporeSigma™ Sterivex™ Sterile Pressure-Driven Devices
Minimize contamination risk to filtered solution by direct-dispensing design. Sterivex Sterile Pressure-Driven Devices are ready-to-use units that filter solutions with volumes of 100 to 2000mL. Available in a variety of fittings, pore sizes and membrane types.
$161.80 - $505.50
Specifications
Color | Clear |
---|---|
Connectors | Female luer lok with upstream vent |
Disposable | Single-use |
Sterility | Sterile |
Filtration Area | 10 cm2 |
Catalog Number | Mfr. No. | Pore Size | Material (Membrane) | Outlet Fitting | No. per Pack | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Material (Membrane) | Outlet Fitting | No. per Pack | Price | Quantity & Availability | ||
SVGP01015
|
MilliporeSigma™
SVGP01015 |
0.22 μm | PES | Male nipple | 15/Pk. |
Each for $221.50
|
|
||
SVGPB1010
|
MilliporeSigma™
SVGPB1010 |
0.22 μm | PES | Filling bell | 10/Pk. |
Each for $164.90
|
|
||
SVGP01050
|
MilliporeSigma™
SVGP01050 |
0.22 μm | PES | Male nipple | 50/Pk. |
Each for $477.50
|
|
||
SVGPL10RC
|
MilliporeSigma™
SVGPL10RC |
0.22 μm | PES | Male luer lok | 15/Pk. |
Pack of 15 for $253.50
|
|
||
SVHV010RS
|
MilliporeSigma™
SVHV010RS |
0.45 μm | PVDF | Male nipple | 50/Pk. |
Pack of 50 for $505.50
|
|
||
SVGV010RS
|
MilliporeSigma™
SVGV010RS |
0.22 μm | PVDF | Male nipple | 50/Pk. |
Pack of 50 for $482.50
|
|
||
SVHV01015
|
MilliporeSigma™
SVHV01015 |
0.45 μm | PVDF | Male nipple | 15/Pk. |
Pack of 15 for $233.50
|
|
||
SVGV01015
|
MilliporeSigma™
SVGV01015 |
0.22 μm | PVDF | Male nipple | 15/Pk. |
Pack of 15 for $248.00
|
|
||
SVGVB1010
|
MilliporeSigma™
SVGVB1010 |
0.22 μm | PVDF | Filling bell | 10/Pk. |
Pack of 10 for $161.80
|
|
||
SVGVL10RC
|
MilliporeSigma™
SVGVL10RC |
0.22 μm | PVDF | Male luer lok | 15/Pk. |
Pack of 15 for $251.00
|
|
||
SVHVB1010
|
MilliporeSigma™
SVHVB1010 |
0.45 μm | PVDF | Filling bell | 10/Pk. |
Pack of 10 for $170.10
|
|
||
SVHVL10RC
|
MilliporeSigma™
SVHVL10RC |
0.45 μm | PVDF | Male luer lok | 15/Pk. |
Pack of 15 for $253.50
|
|
||
Description
Minimize contamination risk to filtered solution by direct-dispensing design. Sterivex Sterile Pressure-Driven Devices are ready-to-use units that filter solutions with volumes of 100 to 2000mL. Available in a variety of fittings, pore sizes and membrane types.
- Sterivex filter units work with syringes, peristaltic pumps or pressure vessels. They eliminate the risk of contamination associated with decanting or pipetting.
- Sterivex units are designed to dispense into virtually any storage container. The devices are available with different outlet connections (see table).
- Sterivex filter units can process volumes up to 1,000 mL or 2,000 mL, depending on the type of membrane used in the device. A choice of three different membranes is provided.
- Filter-sterilize volumes up to 1,000 mL or 2,000 mL, using Sterivex™ pressure-driven filter units (capacity depends on the type of membrane used in the device). With three different membrane options and a variety of outlet connection options, Sterivex™ filters can fit virtually any workflow and dispense into nearly any storage container. Sterivex™ filters work with syringes, peristaltic pumps or pressure vessels and eliminate the risk of contamination associated with decanting or pipetting.
Features & Benefits:
- Works with a variety of pressure devices including syringes, perstaltic pumps and pressure vessels
- Comes in multiple membrane types to suit your application needs with capacities up to 2 L
Applications:
Sterilization of Aqueous Solutions
Specifications
Clear | |
Single-use | |
10 cm2 | |
Fast flow andlow protein binding sterilization of aq. Soln. | |
45°C |
Female luer lok with upstream vent | |
Sterile | |
≥650 mL/min (at 0.7 bar 25°C) | |
113°F | |
Bottle Top Filter |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.