Learn More
MilliporeSigma™ Swinnex™ Filter Holders
Ultraclean or sterilize small volumes of liquids dispensed by syringe
Manufacturer:  MilliporeSigma™ SX0001300
 View more versions of this product
Catalog No. SX0001300
Add to cart
Description
Description
  • 13, 25 and 47mm holders
  • Connect Swinnex 47 with tubing to pressurized reservoir or automatic filling machine for filtering larger volumes
  • 13 and 25mm: polypropylene with silicone gasket
  • 47mm: polypropylene with silicone O-rings
  • Sterilize with a filter in place
Specifications
Specifications
Polypropylene, Silicone
13mm
Documents
Accessories