EMD Millipore™ Thrombin, Human Plasma, Protein Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: EMD Millipore 6051901000U

 View more versions of this product

Catalog No. 60-519-01000U

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Preparation Method Aqueous Buffers or H2O (1mg/mL)
    Purification Method Complete activation from homogeneous prothrombin by SDS-PAGE
    Quantity 1000U
    Immunogen Thrombin
    Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 2 months at −70°C.
    Regulatory Status RUO
    Format Lyophilized
    Formulation Lyophilized from 200mM NaCl, 50mM citrate buffer, 0.1% PEG-8000, pH 6.5. Contains BSA as a stabilizer.
    Host Species Human plasma
    Recombinant No

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit