Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Thrombin, Immobilized, Human Plasma, Protein
Click to view available options
Quantity:
1 mL
Description
Native thrombin from human plasma, bound to agarose beads.

Specifications
Specifications
Concentration | Concentration on Label |
Formulation | In 100mM NaCl, 20mM Tris-HCl, ≤0.1% sodium azide, pH 7.4. |
Quantity | 1 mL |
Immunogen | Thrombin |
Regulatory Status | RUO |
Species | Human |
Recombinant | Native |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction