Learn More
MilliporeSigma™ Ultrafiltration Membranes - Amicon™ XM Polyacrylonitrile/PVC Copolymer Filters
High-recovery membranes for stirred cells
Manufacturer:  MilliporeSigma™ 14342
 View more versions of this product
Catalog No. 14342
Add to cart
Description
Description
  • Regenerated cellulose membranes
  • For concentration of hydrophobic samples or to desalt extremely dilute solutions
  • Hydrophilic, tight microstructure assures high retention with low adsorption of protein, DNA, or other macromolecules
†43mm nominal diameter.
‡62mm nominal diameter.
Specifications
Specifications
2.99 in.
Polyacrylonitrile/PVC Co-polymer
High recovery membranes for stirred cells
10 Pack
7.6cm
300000 NMWL
Ultrafiltration Disc