Learn More
MilliporeSigma™ Ultrafiltration Membranes - BioMax™ PB21
High-recovery membranes for stirred cells
Manufacturer:  MilliporeSigma™ PBTK06210
 View more versions of this product
Catalog No. PBTK06210
Add to cart
Description
Description
  • High-flow, polyethersulfone (PES) membranes
  • Use to concentrate faster or to concentrate and desalt more concentrated samples
  • Open microstructure provides extremely fast separation capability
  • Recommended for serum, plasma, or conditioned tissue culture media samples
Specifications
Specifications
2.5 in.
PBTK
Ultrafiltration
30kDa
Concentrate faster or to concentrate and desalt more concentrated samples
10 Pack
6.35cm
Polyethersulfone
28.7cm2
Serum, plasma or conditioned tissue culture media
Ultrafiltration Membrane