Learn More
EMD Millipore Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
Manufacturer:  EMD Millipore 13622
 View more versions of this product
Catalog No. 13622
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
1.75 in.
YM-10
13.4cm2
10000 NMWL
Ultrafiltration Disc
4.45cm
Adsorptive
Regenerated Cellulose
Protein purification, protein concentration, protein desalting
10 Pack