EMD Millipore Ultrafiltration Membranes - YM

High-recovery membranes for stirred cells

Manufacturer: EMD Millipore 13642

 View more versions of this product

Catalog No. 13642

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Quantity 10 Pack
    Diameter (Metric) Outside 7.6cm
    Material Regenerated Cellulose
    Molecular Weight 10000 NMWL
    Product Type Ultrafiltration Disc
    Diameter (English) Outside 2.99 in.
    Filter Code YM-10
    Filter Type Adsorptive
    Filtration Area 41.8cm2
    For Use With (Application) Protein purification, protein concentration, protein desalting

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit