Learn More
EMD Millipore Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
Manufacturer:  EMD Millipore 13651
 View more versions of this product
Catalog No. 13651
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
3.54 in.
YM-10
63cm2
10000 NMWL
Ultrafiltration Disc
9cm
Adsorptive
Regenerated Cellulose
Protein purification, protein concentration, protein desalting
5 Pack