Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Universal pH Indicator Papers, pH 1 to 14

Click to view available options
Quantity:
3 Rolls
6 Rolls
Description
- High-quality filter paper impregnated with indicator solutions
- M1102320001 are replacement rolls for M1109620003
- Color scale: graduated by 1 pH units
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
pH Range | 1 to 14 |
For Use With (Application) | Environmental analysis and industrial in-process controls, calibration and checking of the products by certified buffer solutions, rapid on-the-spot pH determination |
Graduations | 1-2-3-4-5-6-7-8-9-10-11-12-13-14 |
Quantity | 3 Rolls |
Storage Requirements | 10°C to 25°C |
Type | Universal pH Indicator Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction