Learn More
MilliporeSigma™ Upstate™ NGF 7S Mouse Protein
The mouse NGF 7S Protein( or nerve growth factor 7S) is available in a 100μg format.
Manufacturer:  MilliporeSigma™ 01170
Catalog No. 01-170MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This gene is a member of the NGF-beta family and encodes a secreted protein which homodimerizes and is incorporated into a larger complex. This protein has nerve growth stimulating activity and the complex is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Mutations in this gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of this gene's expression is associated with allergic rhinitis.
Specifications
Specifications
100μg
Male mouse submaxillary glands
Cell culture