Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Vasoactive Intestinal Peptide, Human, Porcine, and Rat, Calbiochem
Supplier: MilliporeSigma™ 052321011MG
Description
Acts as a mitogenic factor for embryonic neurons in the sympathetic nervous system. Has powerful hypotensive and vasodilatory effects. Behaves either as a circulating hormone or as a neurotransmitter. Stimulates cAMP production in rat peritoneal macrophages. Inhibits the growth of human small-cell lung carcinoma (SCLC) both in vitro and in vivo, suggesting that it may be useful as a less toxic alternative to conventional chemotherapeutic agents in the treatment of SCLC. Nitric oxide stimulates the release of this peptide.Specifications
Human, Porcine, Rat | |
≥95% | |
White to Off-white | |
Lyophilized Solid | |
Vasoactive Intestinal Peptide | |
RUO - research use only |
HPLC | |
40077-57-4 | |
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. | |
Supplied as a trifluoroacetate salt. | |
1 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title