Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EME2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543695
Description
Peptide sequence: TTARPHLAVI GLDAYLWSRQ HVSRGTQQPE SPKVAGAEVA VSWPEVEEVR Sequence homology: Human: 100%.
EME2 forms a heterodimer with MUS81 that functions as an XPF-type flap/fork endonuclease in DNA repair.
Specifications
EME2 | |
Polyclonal | |
Unconjugated | |
EME2 | |
EME2; essential meiotic endonuclease 1 homolog 2; essential meiotic structure-specific endonuclease subunit 2; gs125; homolog of yeast EME1 endonuclease 2; probable crossover junction endonuclease EME2; SLX2 structure-specific endonuclease subunit homolog B; SLX2B | |
Rabbit | |
Affinity chromatography | |
RUO | |
197342 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
A4GXA9 | |
EME2 | |
Synthetic peptide directed towards the middle region of human EME2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction