Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EME2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen EME2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095988
View Documents
Novus Biologicals
NBP304534100UL
100 μg
Each for $320.71
Add to cart
 
NB095989
View Documents
Novus Biologicals
NBP30453420UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

EME2 Polyclonal antibody specifically detects EME2 in Human samples. It is validated for Western Blot
Specifications

Specifications

EME2
Unconjugated
Rabbit
Human
EC 3.1.22, Essential Meiotic Endonuclease 1 Homolog 2, Essential Meiotic Endonuclease 1 Homolog 2 (S. Pombe), Essential Meiotic Structure-Specific Endonuclease Subunit 2, gs125, Homolog Of Yeast EME1 Endonuclease 2, Probable Crossover Junction Endonuclease EME2, SLX2 Structure-Specific Endonuclease Subunit Homolog B, SLX2 Structure-Specific Endonuclease Subunit Homolog B (S. Cerevisiae), SLX2B
Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human EME2 (NP_001244299.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
197342
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit