Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Emetine Hydrochloride, USP, 98-101.5%, Spectrum™

Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle

E1069, 316-42-7, C29H42Cl2N2O4

Spectrum™ Emetine Hydrochloride, USP is a drug used as both an anti-protozoal and to induce vomiting. All Spectrum Chemical USP products are packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Quantity 100 g
Ignition Residue 0.002
CAS 316-42-7
CAS Max % 1
Molecular Formula C29H42Cl2N2O4
UN Number UN1544
InChI Key JROGBPMEKVAPEH-GXGBFOEMSA-N
SMILES [H+].[H+].[Cl-].[Cl-].CC[C@H]1CN2CCC3=CC(OC)=C(OC)C=C3[C@@H]2C[C@@H]1C[C@H]1NCCC2=CC(OC)=C(OC)C=C12
IUPAC Name dihydrogen (1R)-1-{[(2S,3R,11bS)-3-ethyl-9,10-dimethoxy-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinolin-2-yl]methyl}-6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline dichloride
Molecular Weight (g/mol) 553.57
Percent Purity 98 to 101.5%
Show More Show Less