Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMG1 Mouse anti-Human, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI1B8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271601APCCY7

Catalog No. N271601APC7


Add to cart

Description

Description

EMG1 Monoclonal antibody specifically detects EMG1 in Human, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

EMG1
OTI1B8
Flow Cytometry
Affinity Purified, Conjugated
EMG1
Full length recombinant protein of human EMG1 (NP_006322) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
18S rRNA (pseudouridine-N1-)-methyltransferase NEP1, BWCNS, C2F18S rRNA Psi1248 methyltransferase, EMG1 nucleolar protein homolog (S. cerevisiae), essential for mitotic growth 1, FLJ60792, Grcc2f, NEP1EC 2.1.1.-, Nucleolar protein EMG1 homolog, probable ribosome biogenesis protein NEP1, Protein C2f, Ribosome biogenesis protein NEP1
Mouse
IgG2b
0.1 mL
Primary
10436.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit