Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMG1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548126

Catalog No. PA548126

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

EMG1 involved in 40S ribosomal subunit biogenesis. It seems to play a role a methylation reaction in pre-rRNA processing.
Specifications

Specifications

EMG1
Polyclonal
PBS with 0.09% sodium azide
Q92979
Ribosomal RNA small subunit methyltransferase NEP1, 211-, 18S rRNA (pseudouridine(1248)-N1)-methyltransferase, 18S rRNA Psi1248 methyltransferase, Nucleolar protein EMG1 homolog, Protein C2f, Ribosome biogenesis protein NEP1, EMG1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10436
Flow Cytometry, Western Blot
Unconjugated
EMG1
Liquid
EMG1
KLH conjugated synthetic peptide between 104-133 amino acids from the Central region of human EMG1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit