Learn More
EML4 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 227091AP
Catalog No. 50-173-3930
Add to cart
Description
Description
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
Specifications
Specifications
EML4
0.44 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9HC35
C2orf2, ELP120, EMAP 4, EMAPL4, EML4, Ropp 120, ROPP120
Rabbit
IgG
150 μL
-20°C
Primary
27436
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
EML4
Liquid
EML4
EML4 Fusion Protein Ag18463
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human