Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Emmet Iris Hook, Sklar, 12 in., 3mm, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902124
Sklar Black Emmet Iris Hook, Sklar, 12 in., 3mm, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Maxi Iris Hook, is most commonly used in OB/GYN procedures involving the uterus. The hook is at the end of a long, narrow handle, often 9 or 12 inches in length. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. This product is straight with a 3mm hook tip and an overall length of 12 inches.
Catalog No. 50-143-6549