Learn More
Sklar InstrumentsSupplier Diversity Partner Emmett Hook, Sklar, 9 in. Emmett Hook, Style #1, Curved, Pointed, Solid Square, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 908301
Emmett Hook, Sklar, 9 in. Emmett Hook, Style #1, Curved, Pointed, Solid Square, Stainless Steel. Sklar's Emmett Tenaculum Hook is used to hold the uterus and cervix during OB/GYN procedures such as to insert an intrauterine device (IUD or coil). The hook comes in a variety of styles. This product is style #1 which is a small half curved hook with an overall length of 9 inches.
Catalog No. 50-143-6861