Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Emmett Tenaculum Hook, Sklar, #3, Angled (90 Degrees), Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 902173
Sklar Black Emmett Tenaculum Hook, Sklar, #3, Angled (90 Degrees), Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Emmett Tenaculum Hook is used to hold the uterus and cervix during OB/GYN laser procedures. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. The hook comes in a variety of styles. This product is style #3 which is a 90 degree angled hook with an overall length of 9 inches.
Catalog No. 50-143-6557