Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Mouse anti-Human, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI9H5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270278APCCY7

Catalog No. N270278APC7


Add to cart

Description

Description

EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

EMMPRIN/CD147
OTI9H5
Flow Cytometry
Affinity Purified, Conjugated
BSG
Full length recombinant protein of human EMMPRIN/CD147 (NP_001719) produced in HEK293T cells.
Immunogen affinity purified
Cancer, Signal Transduction, Vision
Primary
682.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit