Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EMMPRIN/CD147 Mouse anti-Human, Rat, Canine, Primate, Clone: OTI9H5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270278
Description
EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Human, Rat, Canine, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF.Specifications
EMMPRIN/CD147 | |
OTI9H5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
Mouse | |
IgG2b | |
Immunogen affinity purified | |
Cancer, Signal Transduction, Vision | |
Primary | |
682 | |
Canine, Human, Primate, Rat |
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
LYOPH | |
Western Blot 1:1500-3000, Flow Cytometry 1:100, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready | |
Affinity Purified, Lyophilized | |
BSG | |
Full length human recombinant protein of human BSG(NP_001719) produced in HEK293T cell. | |
40.2 kDa | |
100 μg | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title