Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Rabbit anti-Human, DyLight 650, Clone: 118, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289319C

Catalog No. NB066243


Add to cart

Description

Description

EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Human samples. It is validated for Sandwich ELISA Detection.
Specifications

Specifications

EMMPRIN/CD147
118
Sandwich ELISA Detection
Purified
Rabbit
IgG
0.1 mL
Cancer, Signal Transduction, Vision
Primary
682
ELISA
DyLight 650
50mM Sodium Borate
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
This antibody was obtained from a rabbit immunized with purified, recombinant Human EMMPRIN/CD147 (NP_940991.1; Met1-His205).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit