Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen EMMPRIN/CD147
Applications Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092629
View Documents
Novus Biologicals
NBP303732100UL
100 μg Each for $320.71
Add to cart
 
NB092630
View Documents
Novus Biologicals
NBP30373220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

EMMPRIN/CD147 Polyclonal antibody specifically detects EMMPRIN/CD147 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, KnockDown
Specifications

Specifications

EMMPRIN/CD147
Unconjugated
Rabbit
Cancer, Signal Transduction, Vision
PBS with 50% glycerol, pH7.3.
682
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot
Purified
RUO
Human, Mouse, Rat
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Recombinant fusion protein containing a sequence corresponding to amino acids 45-200 of human EMMPRIN/CD147 (NP_940991.1).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit