Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EMMPRIN/CD147 Rabbit anti-Mouse, DyLight 405, Clone: 41, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP290540V

Catalog No. NB039954


Add to cart

Description

Description

EMMPRIN/CD147 Monoclonal antibody specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for ELISA, Immunofluorescence
Specifications

Specifications

EMMPRIN/CD147
41
ELISA, Immunocytochemistry/Immunofluorescence
Purified
Rabbit
IgG
0.1 mL
Cancer, Signal Transduction, Vision
Primary
682
ELISA, Immunofluorescence
DyLight 405
50mM Sodium Borate
5F7, basigin, basigin (Ok blood group), CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor
Recombinant Mouse EMMPRIN/CD147 protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit