Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ Emperor™ ME108 Chemical Protection Gloves

Ansell™ Emperor™ ME108 Chemical Protection Gloves provide good mechanical and chemical protection in heavy duty enrivonments

$217.00

Specifications

Certifications/Compliance REACH Compliant
Certifications/Compliance (Europe) EN420:2003 + A1:2009; Category lll
Color Black
Exterior Finish Smooth
Glove Cuff Style Beaded
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-010-400
View Documents
Ansell™
10463
9.5
Case of 12 for $217.00
Add to cart
 
19-010-401
View Documents
Ansell™
10464
10.5
Case of 12 for $217.00
Add to cart
 
Description

Description

  • Extra heavy duty chemical protection glove
  • Beaded cuff for easy donning
  • 610mm long glove length
  • High resistance to water-based chemicals
  • Good flexibility and elasticity preventing tears
  • Chlorinated to harden and cleanse the surface of the glove
  • AQL 0.65
Specifications

Specifications

REACH Compliant
Black
Beaded
24 in.
Emperor™ ME108
1 pair/bag; 12 bags/carton
1.5 mm
EN420:2003 + A1:2009; Category lll
Smooth
No
Rubber Latex
Glove
Emperor™ ME108
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit