Learn More
CDS Analytical™ Empore™ C18 StageTips
C18 bonded silica StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
Supplier: CDS Analytical™ 70201910013
This item is not returnable.
View return policy
Description
The CDS™ Empore™ StageTips C18 bonded silica solid phase extraction micropipette tips have two layers for increased capacity.
- Combines good retention capacity with high recoveries
- Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
12 μm | |
C18 (Octadecyl, ODS) | |
960 Cs. |
Empore | |
200 μL |