Learn More
CDS Analytical™ Empore™ C18 StageTips
SDP
C18 bonded silica StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
$209.00 - $1463.00
Specifications
Particle Size 12 μm
Product Line Empore
Sorbent C18 (Octadecyl, ODS)
Volume (Metric) 200 μL
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
13-110-055
SDP
View Documents
CDS Analytical™
70201910013EA
96 Pk.
Pack of 96 for $209.00
Only null left
Add to Cart
13-110-056
SDP
View Documents
CDS Analytical™
70201910013
960 Cs.
Case of 960 for $1,463.00
Only null left
Add to Cart
Description
Description
The CDS™ Empore™ StageTips C18 bonded silica solid phase extraction micropipette tips have two layers for increased capacity.
  • Combines good retention capacity with high recoveries
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
12 μm
Empore
C18 (Octadecyl, ODS)
200 μL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.