Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One™ Empty Rack for Sapphire Pipette Tips

For use with Saphire™ universal pipette tips.

Manufacturer:  Greiner Bio-One™ 970250

 View more versions of this product

Catalog No. 07-000-972

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Color coded for ease of use
  • Made of polypropylene
  • Unfilled
  • Stackable
  • Lids are unhinged for smooth opening/closing action
  • Reversible lid to enable opening/closing action
Specifications

Specifications

Rack
Blue
Nonsterile
1000 and 1250μL
Polypropylene
Empty rack
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit