Learn More
Seca Corp 644 EMR-validated handrail scale with extra-large platform
Supplier:  Seca Corp 6441321108
Encompass_Preferred
Equipped with an extremely low-level, easy-access platform, a stable handrail, the seca 644 multifunctional scale ensures safe and comfortable weighing of heavy patients. The integrated handrail provides the patient with support throughout the weighing process. The pre-TARE can be used to automatically deduct a stored weight (such as a chair) to determine net weight. • Safe weighing due to sturdy handrail • Large non-slip platform with 800 pounds capacity • Swiveling display • BMI calculator and damping system • Mobile with transport wheels • EMR-validated: transmit measured data directly to any Electronic Medical Record (EMR) system • Capacity: 800 lbs / 360 kg • Graduation: 0.1 lbs / 50 g • Functions: TARE, pre-TARE, HOLD, auto-HOLD, BMI, auto-CLEAR, kg/lbs switch-over, damping, automatic switch-off • Optional: measuring rod seca 223
Catalog No. NC1798770