Learn More
Abnova™ EMR3 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00084658T01
Catalog No. 89-943-245
Add to cart
Specifications
Specifications
Western Blot
293T
Theoretical M.W. 72.6kDa
Transient overexpression cell lysate was tested with anti-EMR3 antibody (Mfr. No. H00084658-B01) byWesternBlots.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
EMR3 293T Cell Transient Overexpression Lysate(Denatured)
pCMV-EMR3 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.