Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ENC1 Polyclonal Antibody
Chicken Polyclonal Antibody
Supplier: Invitrogen™ PA520936
Description
A suggested positive control is mouse kidney tissue lysate. PA5-20936 can be used with blocking peptide PEP-1050.
The ectoderm-neural cortex-1 (ENC-1) protein is an early and highly specific marker of neural induction in vertebrates. It is a kelch family related protein that functions as an actin-binding protein and has been suggested to be involved in the organization of the actin cytoskeleton during neural fate specification and development of the nervous system. ENC-1 has also been shown to be required for adipocyte differentiation when cytoskeletal reorganization and cell shape change from fibroblastic preadipocytes to spherical adipocytes occur.
Specifications
ENC1 | |
Polyclonal | |
Unconjugated | |
ENC1 | |
CCL28; ectodermal-neural cortex (with BTB-like domain); ectodermal-neural cortex 1; ectodermal-neural cortex 1 (with BTB domain); ectodermal-neural cortex 1 (with BTB-like domain); Ectoderm-neural cortex protein 1; Enc1; ENC-1; FLJ39259; kelch-like 35; kelch-like 37; kelch-like family member 37; kelch-like protein 37; KLHL35; KLHL37; NRPB; Nuclear matrix protein NRP/B; nuclear restrict protein in brain; nuclear restricted protein, BTB domain-like (brain); p53-induced gene 10 protein; PIG10; si:dkey-35i22.1; TP53I10; tumor protein p53 inducible protein 10 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
13803, 294674, 8507 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O14682, O35709 | |
ENC1 | |
A 13 amino acid peptide near the center of human ENC-1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction