Learn More
Encon 160 Series Goggles
Impact resistant lens provides distortion-free 160° field of vision
Manufacturer:  ENCON SAFETY PRODUCTS 5068009
 View more versions of this product
Catalog No. 19-130-1943
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Safety goggles
Anti-fog,Anti-UV
Black
Black Elastic Strap
ENFOG
Clear
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
PVC
99.90%