Encon 160 Series Goggles
Impact resistant lens provides distortion-free 160° field of vision
Manufacturer: ENCON SAFETY PRODUCTS 05068004
 View more versions of this product
Catalog No. 19-013973
  Check Availability
Add to Cart
    Specifications
    Lens Tint Clear
    Material Polycarbonate
    Surface Coating Enfog Antifog
    Lens Coating ENFOG
    Type Safety Goggles
    Certifications/Compliance ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
    Color Clear
    Frame Color Clear
    Frame Material PVC
  • Description & Specifications
Related Products
Product Content Feedback