Learn More
Encon™ 500 Series Goggles
Aspheric correction allows distortion-free optics
Manufacturer:  ENCON SAFETY PRODUCTS 05057204
 View more versions of this product
Catalog No. 19-014-012
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Encon™ 500 Series 501R Safety Goggles
Clear
PVC
Clear
Uncoated
Indirect
Clear
ANSI Z87.1-2010, CSA Z94.3-2007
Gray
Black Neoprene Strap, Indirect Venting, SV2 Side Vent
Polycarbonate
99.90%
Uncoated