Encon™ 500 Series Goggles
Aspheric correction allows distortion-free optics
Manufacturer: ENCON SAFETY PRODUCTS 05058204
 View more versions of this product
Catalog No. 19-014019
  Check Availability
Add to Cart
    Specifications
    Includes Black Neoprene Strap, Indirect Venting, SV2 Side Vent
    Lens Tint Clear
    Surface Coating Uncoated
    Lens Coating ENFOG™ coating
    Lens Type Clear, with Enfog coating
    Type Encon™ 500 Series 503R Safety Goggles
    Certifications/Compliance ANSI Z87.1-2010, CSA Z94.3-2007
    Color Clear
    Frame Color Gray
    Frame Material Plastic
  • Description & Specifications
Related Products
Product Content Feedback