Learn More
Encon™ 500 Series Goggles
Aspheric correction allows distortion-free optics
Manufacturer:  Encon™ 05058202
 View more versions of this product
Catalog No. 19-062076
Add to cart
Description
Description
  • Impact-resistant polycarbonate lens provides 99.99% UV absorption up to 380nm
  • Optional Enfog™ antifog coating
  • Lightweight PVC gray plastic frame with universal fit
  • Indirect ventilation system with labyrinth design protects eyes from liquid, dust, and airborne particulates; allows cooling, continuous airflow
  • Forward tilt allows superior field of vision for wearer working at benchtop
  • Black neoprene strap
Order Info
Cat. No. 19-048-483 includes quick-draw strap retainer.
Certifications
CSA Z94.3-2007 standards.
Compliance
Meet ANSI Z87.1-2010-High Impact
Specifications
Specifications
Encon™ 500 Series 503RC Safety Goggles
Anti-fog
ANSI Z87.1-2010, CSA Z94.3-2007
Black Neoprene Strap, Felt Binding, Indirect Venting, SV2 Side Vent
ENFOG™ coating
Clear
Safety
Gray
Vinyl