Learn More
Encon™ Encon™ Aquarion™ Self-Contained Eyewash
For use on any wall, tight hallway, column, shelf or cart
Manufacturer:  Encon™ AQ100
Catalog No. 19-811-191
Add to cart
Description
Description
  • Sealed cartridge gravity feed operating device
  • Yellow ABS plastic outer body for protection of fluid bags and internal cavity of station
  • Side windows for inspection of fluid bags without removal of the cover
  • Polycarbonate backplate
  • Mounting slots to screw to wall studs
  • Retracting tray for protecting nozzels with single pull down motion
  • Includes usage indicator
  • Plastic heads connect to retracting tray; non-injurious, dual stream, of water at 0.4gpm (1.5 lpm)
Certifications
ANSI 358.1, CSA
Specifications
Specifications
Portable Eyewash
ANSI Z358.1, CSA
ABS Plastic outer body, retracting tray, side windows, sealed cartridge