Learn More
Combination Eye/Facewash and Shower Units with Galvanized Steel Piping
Encon Combination Eye/Facewash and Shower Units provides workers with quick access to eye and face flushing in case of an emergency.
Manufacturer:  ENCON SAFETY PRODUCTS 01050290
 View more versions of this product
Catalog No. 19-021729
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Combination shower/eye/facewash unit
ANSI Z358.1-2009, CSA Z94.3-2007
Yellow ABS Plastic shower head, ABS Plastic dual stream eyewash head, wye, ABS Plastic yellow bowl, Stainless Steel bead chains, self adjusting regulators, foot treadle, Galvanized pipe
30 to 70psi
Stainless Steel push plate and pull rod
3.2 - 30 gpm
1.25 in. NPT