Learn More
Combination Eye/Facewash and Shower Units with Galvanized Steel Piping
Encon Combination Eye/Facewash and Shower Units provides workers with quick access to eye and face flushing in case of an emergency.
Manufacturer:  ENCON SAFETY PRODUCTS 01050230
 View more versions of this product
Catalog No. 18-999-339A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Combination shower/eyewash unit
ANSI Z358.1-2009, CSA Z94.3-2007
Yellow ABS plastic shower head, acetal plastic dual stream head, BCP wye, integral hinged covers, stainless steel bowl, chrome plated bronze stay open ball valves, self adjusting regulators, galvanized pipe, stainless steel bead chain, yellow actuation graphic
30 to 70psi
Stainless Steel push plate and pull rod
2.4 - 30 gpm
1.25 in. NPT