Encon™ Combination Eye/Facewash and Shower Units with Galvanized Steel Piping
Encon Combination Eye/Facewash and Shower Units provides workers with quick access to eye and face flushing in case of an emergency.
Manufacturer: Encon™ 01050221
 View more versions of this product
Catalog No. 19-021724
  Check Availability
Add to Cart
    Specifications
    Product Type Combination shower/eye/facewash unit
    Flow Rate 8 - 30 gpm
    Includes Yellow ABS Plastic shower head, ABS Plastic facewash head, wye, Chrome Plated Bronze stay open ball valves, self adjusting regulators
    Pressure Range 30 to 70psi
    Activation Stainless Steel push plate and pull rod
    Certifications/Compliance ANSI Z358.1-2009, CSA Z94.3-2007
  • Description & Specifications
Related Products
Product Content Feedback