Learn More
Combination Eye/Facewash and Shower Units with Galvanized Steel Piping
Encon Combination Eye/Facewash and Shower Units provides workers with quick access to eye and face flushing in case of an emergency.
Manufacturer:  ENCON SAFETY PRODUCTS 01050234
 View more versions of this product
Catalog No. 19-021726
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Combination shower/eye/facewash unit
ANSI Z358.1-2009, CSA Z94.3-2007
Yellow ABS Plastic shower head, ABS Plastic dual stream eyewash/facewash head, Stainless Steel bowl, Chrome Plated Bronze stay open ball valves, self adjusting regulators, BCP wye, Galvanized pipe
30 to 70psi
Stainless Steel push plate and pull rod
8 - 30 gpm
1.25 in. NPT