Learn More
Encon™ Counter-Mount Eyewash/Drench Hose Station
Outfit your workspace and protect your employees with the Encon Counter-Mount Eyewash/Drench Hose Station.
Manufacturer:  Encon™ 01090012
 View more versions of this product
Catalog No. 19-035-064
  Check Availability
Add to cart
Specifications
Description
  • Installed in sink/countertop area, this handheld dual-stream hose features solvent-resistant acetal, dual-stream heads with integral hinged covers
  • Heads are 8 1/2 in. (21.6cm) from countertop, mounted on chrome-plated brass “T” piping
  • Retractable 8 ft. long (2.4m) synthetic rubber hose attaches directly to counter-mount station and is stored under the countertop
  • To operate, simply pull the unit from its countertop base. Squeeze lever handle to open the 3/8 in. (1cm) chrome-plated brass self-closing lockable valve and release spray. Slip the locking mechanism over the handle to secure valve in “open” position
  • Self-adjusting regulators ensure a constant, even flow of 2.4 gpm (9.1L/min.) under varying hydraulic conditions1
  • Includes universal language sign, sink/countrtop mounting bracket, and 3/8 in
  • NPT connection (1/2 in. supply line recommended).
Description & Specifications
Specifications
ANSI Z358.1-2004
CP Bronze value, Acetal head with hinged cover, Synthetic Rubber hose, sink/countertop mounting bracket
0.939cm
2.44m
Dual eyewash/drench hose station
2.4 gpm
0.37 in.
8 ft.
Countertop