Encon Countertop Eyewashes
Manufacturer: ENCON SAFETY PRODUCTS 01040123
 View more versions of this product
Catalog No. 18-999-341B
  Check Availability
Add to Cart
    Specifications
    Product Type Eyewash
    Flow Rate 2.4 gpm
    For Use With (Application) Laboratory use
    Includes Acetal eyewash head, hinged cover, vertical valve, right hand actuation, self adjusting regulator, Chrome Plated Bronze ball valve and ball
    Activation Stainless Steel push plate
    Certifications/Compliance ANSI Z358.1-1998
  • Description & Specifications
Related Products
Product Content Feedback